4-Methylpyrocatechol 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4-Methylbenzene-1,2-diol, 4-Methylcatechol
126510050 126510250 126511000 126515000
ACRO126510050EA 375 SEK
ACRO126510050 ACRO126510250 ACRO126511000 ACRO126515000
4-Methylpyrocatechol 98%
4-Methylpyrocatechol
Formel: C₇H₈O₂
MW: 124.14 g/mol
Kokpunkt: 251 °C (760 mmHg)
Smältpunkt: 62…69 °C
Flampunkt: 140 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002205
CAS nummer: 452-86-8
EINECS: 207-214-5
Order Now
Specifikation testresultat
Appearance White to brown Powder and/or chunks
Infrared spectrum Conforms
Melting point 63 to 68 °C
GC ≥97.5 %