6-Methyl-2-thiouracil 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Methiocil, 2-Mercapto-6-methylpyrimidin-4(1H)-one, Methylthiouracil, MZU, Basethyrin, 4-Hydroxy-2-mercapto 6-methylpyrimidine, Thiothymin
121305000 121300025
ACRO121305000EA 1425 SEK
ACRO121305000 ACRO121300025
6-Methyl-2-thiouracil 98%
6-Methyl-2-thiouracil
Formel: C₅H₆N₂OS
MW: 142,18 g/mol
Kokpunkt: 351 °C (1013 hPa)
Smältpunkt: 330 °C (dec.)
Densitet: 1,519 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00006040
CAS nummer: 56-04-2
EINECS: 200-252-3
Merck Index: 12,06210
Order Now
Specifikation testresultat
Appearance White to off-white Powder or crystals and/or chunks
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH ≥96.0 %