Cyanoättiksyra 97%
Leverantör: Thermo Fisher Scientific
Fara
110670010 110670250 110672500 110670050
ACRO110670010EA 2475 SEK
ACRO110670010 ACRO110670250 ACRO110672500 ACRO110670050
Cyanoättiksyra 97%
Cyanoättiksyra
Formel: C₃H₃NO₂
Kokpunkt: 108 °C (15 mmHg)
Smältpunkt: 65…67 °C
Densitet: Min, 1.1 g/cm³ (20 °C)
Flampunkt: 107 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002677
CAS nummer: 372-09-8
EINECS: 206-743-9
UN: 3261
ADR: 8,II
Merck Index: 13,02718
Order Now
Specifikation testresultat
Appearance White to light beige Adhering crystalline powder or crystals
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %
Specific optical rotation -10° to -12° (20°C, 589 nm) (neat)
Titration with HClO4 ≥97.5 %