4-Bromobutyronitrile 97%
Leverantör: Thermo Scientific
|
Varning
|
106800050
106800250
ACRO106800050EA
630
SEK
ACRO106800050
ACRO106800250
4-Bromobutyronitrile 97%
4-Bromobutyronitrile
Formel:
BrH₂CCH₂CH₂CN Kokpunkt: 205 °C (1013 hPa) Densitet: 1,48 g/cm³ Flampunkt: 103 °C |
MDL Number:
MFCD00001971 CAS nummer: 5332-06-9 EINECS: 226-236-6 UN: 3276 ADR: 6.1,III |
Specifikation testresultat
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4770 to 1.4790 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...