3-Benzoylpropionic acid 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Oxo-4-phenylbutanoic acid, 3-Benzoylpropionicacid
105790250 105791000
ACRO105790250EA 1225 SEK
ACRO105790250 ACRO105791000
3-Benzoylpropionic acid 98%
3-Benzoylpropionic acid
Formel: C₁₀H₁₀O₃
MW: 178,19 g/mol
Kokpunkt: 325 °C (1013 hPa)
Smältpunkt: 114…119 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002792
CAS nummer: 2051-95-8
EINECS: 218-135-0
Order Now
Specifikation testresultat
Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 114 to 119 °C
HPLC ≥97.5 %