Benzofuran 99.5%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,3-Benzofuran, Benzo[b]furan, Benzo(b)furan, 1-Benzofuran, Coumarone
105480100 105480500
ACRO105480100EA 1225 SEK
ACRO105480100 ACRO105480500
Benzofuran 99.5%
Benzofuran
Formel: C₈H₆O
MW: 118,14 g/mol
Kokpunkt: 173…175 °C (760 mmHg)
Smältpunkt: < –18 °C
Densitet: 1,07 g/cm³
Flampunkt: 50 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00005847
CAS nummer: 271-89-6
EINECS: 205-982-6
UN: 1993
ADR: 3,III
Merck Index: 13,01089
Order Now
Specifikation testresultat
Appearance Clear yellow to orange-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5655 to 1.5685 (20°C, 589 nm)
GC ≥99.4 %