7-Azaindole 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1H-Pyrrolo[2,3-b]pyridine
105100050 105100010 105100250
ACRO105100050EA 2025 SEK
ACRO105100050 ACRO105100010 ACRO105100250
7-Azaindole 98%
7-Azaindole
Formel: C₇H₆N₂
MW: 118,14 g/mol
Kokpunkt: 270 °C (1004HPA)
Smältpunkt: 103…107 °C
Densitet: 1,301 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00005606
CAS nummer: 271-63-6
EINECS: 205-981-0
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 103 to 107 °C
GC ≥97.5 %