Toluen

Leverantör: ABSOR
Fara

1160 1161
ABSO1160EA 2025 SEK
ABSO1160 ABSO1161
Toluen
Toluen
Formel: C₆H₅CH₃
Kokpunkt: 110,6 °C (1013 hPa)
Smältpunkt: –95 °C
Densitet: 0,867 g/cm³ (20 °C)
Flampunkt: 4 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008512
CAS nummer: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR