2-Propanol

Leverantör: ABSOR
Fara

1068 1069
ABSO1068EA 3450 SEK
ABSO1068 ABSO1069
2-Propanol
2-Propanol
Formel: (CH₃)₂CHOH
Kokpunkt: 82 °C (1013 hPa)
Smältpunkt: –89 °C
Densitet: 0,786 g/cm³ (20 °C)
Flampunkt: 12 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011674
CAS nummer: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR