Hematoxylin, Sigma-Aldrich®
Leverantör: SIGMA ALDRICH MICROSCOPY
Synonyms: DcH-48, Hydroxybrazilin, (6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaol, NSC 2785, Natural Black I, Hydroxybrasilin, (+)-Hematoxylin
M-Clarity™ quality level = MQ200
H3136-100G H3136-25G
H3136-100GEA 5880 SEK
H3136-100G H3136-25G
Hematoxylin, Sigma-Aldrich®
Hematoxylin
Formel: C₁₆H₁₄O₆
MW: 302,28 g/mol
Smältpunkt: 291…293 °C
Storage Temperature: Omgivande
MDL Number: MFCD00078111
CAS nummer: 517-28-2
EINECS: 208-237-3
Merck Index: 13,04654
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a1a11675c83ab3776411bf728e6c1426020b0a4&vid=bd1a3ee5236b13437564745d59df067c9ecb8106&prid=web&clid=SAPDAM