Hematoxylin 85%, ren, certified
Leverantör: Thermo Scientific
Varning
Synonyms: DcH-48, Hydroxybrazilin, (6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaol, NSC 2785, Natural Black I, Hydroxybrasilin, (+)-Hematoxylin
411160250 411161000 411165000
ACRO411160250EA 2200 SEK
ACRO411160250 ACRO411161000 ACRO411165000
Hematoxylin 85%, ren, certified
Hematoxylin
Formel: C₁₆H₁₄O₆
MW: 302,28 g/mol
Smältpunkt: 100…120 °C
Storage Temperature: Omgivande
MDL Number: MFCD00078111
CAS nummer: 517-28-2
EINECS: 208-237-3
Merck Index: 13,04654
Order Now
Specifikation testresultat
Appearance Light beige to brown Powder
Infrared spectrum Conforms
Loss on drying ≤10 % (1 g, 105°C, 3 h)
UV spectrophotometry ≥80 % (On dry basis)
BSC Certification Passes