L (-)-Proline, Millipore®
Leverantör: Merck
Synonyms:
(S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH
M-Clarity™ quality level = MQ100
1.07434.0010
1.07434.0100
1.07434.0500
1.07434.9010
1.07434.0010EA
450
SEK
1.07434.0010
1.07434.0100
1.07434.0500
1.07434.9010
L (-)-Proline, Millipore®
L (-)-Proline
Formel:
C₅H₉NO₂ MW: 115.13 g/mol Kokpunkt: 245 Smältpunkt: 220…222 °C Densitet: 1.35 g/cm³ (25 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00064318 CAS nummer: 147-85-3 EINECS: 205-702-2 Merck Index: 13,07871 |