L-Proline, EMPROVE® ESSENTIAL Ph. Eur., USP suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Synonyms:
(S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH
1.07430.9010
1.07430.1000
1.07430.9010EA
0
SEK
1.07430.9010
1.07430.1000
L-Proline, EMPROVE® ESSENTIAL Ph. Eur., USP suitable for use as excipient, SAFC®
L (-)-Proline
Formel:
C₅H₉NO₂ Kokpunkt: 245 °C (1013 hPa) Smältpunkt: 220…222 °C Densitet: 1,35 g/cm³ (25 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00064318 CAS nummer: 147-85-3 EINECS: 205-702-2 Merck Index: 13,07871 |