D-(+)-Proline ≥99%
Leverantör: Thermo Scientific
Synonyms: H-D-Pro-OH, (R)-(+)-Proline, D-Proline
A10682.03 A10682.06 A10682.14
A10682.03EA 1200 SEK
A10682.03 A10682.06 A10682.14
D-(+)-Proline ≥99%
D-(+)-Proline
Purity: 99% CAS No.: 344-25-2 Molecular Formula: C5H9NO2 Formula Weight: 115.13 Melting Point: 213-215° dec. Beilstein Reference: 80811
Formel: C₅H₉NO₂
MW: 115,13 g/mol
Smältpunkt: 213…215 °C (dec.)
Densitet: 1,36
Storage Temperature: Omgivande
MDL Number: MFCD00064317
CAS nummer: 344-25-2
EINECS: 206-452-7