(R)-(-)-Mandelic acid ≥99%, ee 99%, ChiPros®
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: D(-)-Mandelic acid, (R)-(-)-2-Hydroxy-2-phenylacetic acid, (R)-2-Hydroxy-2-phenylacetic acid, D(-)-2-Hydroxy-2-phenylacetic acid
L19116.14 L19116.06
L19116.14EA 2050 SEK
L19116.14 L19116.06
(R)-(-)-Mandelic acid ≥99%, ee 99%, ChiPros®
(R)-(-)-Mandelic acid
Purity: 99+% CAS No.: 611-71-2 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 131-135° Beilstein Reference: 2691094
Formel: C₈H₈O₃
MW: 152.15 g/mol
Kokpunkt: 322
Smältpunkt: 131…135 °C
Densitet: 1.341 g/cm³ (20 °C)
Flampunkt: >190 °C
Storage Temperature: Omgivande
MDL Number: MFCD00064251
CAS nummer: 611-71-2
EINECS: 210-276-6