2-Methyl-4-(trifluoromethoxy)phenylboronic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Methyl-4-(trifluoromethoxy)benzeneboronic acid
PC7578-250MG PC7578-1G
APOSPC7578-250MGEA 1575 SEK
APOSPC7578-250MG APOSPC7578-1G
2-Methyl-4-(trifluoromethoxy)phenylboronic acid 98%
2-Methyl-4-(trifluoromethoxy)phenylboronic acid
Formel: C₈H₈BF₃O₃
MW: 219.950 g/mol
Smältpunkt: 186…190 °C
Storage Temperature: Kylskåp
MDL Number: MFCD09475816
CAS nummer: 850033-39-5