5-Bromo-2-(cyclopropyl)thiophene-3-carbonitrile 95%
Leverantör: Apollo Scientific
Fara
OR45205-250MG OR45205-1G
APOSOR45205-250MGEA 2575 SEK
APOSOR45205-250MG APOSOR45205-1G
5-Bromo-2-(cyclopropyl)thiophene-3-carbonitrile 95%
5-Bromo-2-(cyclopropyl)thiophene-3-carbonitrile
Formel: C₈H₆BrNS
Storage Temperature: Kylskåp
MDL Number: MFCD32219467
UN: 3276
ADR: 6.1,III