Sodium rel-(1R,2r,3S,4R,5s,6S)-cyclohexane-1,2,3,4,5,6-hexayl hexakis(phosphate) 96%
Leverantör: Apollo Scientific
Varning
Synonyms: Sodium phytate, Dodecasodium (2,3,4,5,6-pentaphosphonatooxycyclohexyl) phosphate
OR71104-100G OR71104-500G OR71104-1KG
APOSOR71104-100GEA 775 SEK
APOSOR71104-100G APOSOR71104-500G APOSOR71104-1KG
Sodium rel-(1R,2r,3S,4R,5s,6S)-cyclohexane-1,2,3,4,5,6-hexayl hexakis(phosphate) 96%
Sodium rel-(1R,2r,3S,4R,5s,6S)-cyclohexane-1,2,3,4,5,6-hexayl hexakis(phosphate)
Formel: C₆H₆Na₁₂O₂₄P₆
MW: 923,81 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00082310
CAS nummer: 14306-25-3