(S)-2-Amino-2-(2-(trifluoromethyl)phenyl)ethan-1-ol hydrochloride 95%
Leverantör: Apollo Scientific
Varning
PC57466-100MG PC57466-250MG PC57466-1G
APOSPC57466-100MGEA 3600 SEK
APOSPC57466-100MG APOSPC57466-250MG APOSPC57466-1G
(S)-2-Amino-2-(2-(trifluoromethyl)phenyl)ethan-1-ol hydrochloride 95%
(S)-2-Amino-2-(2-(trifluoromethyl)phenyl)ethan-1-ol hydrochloride
Formel: C₉H₁₁ClF₃NO
MW: 241.6380 g/mol
MDL Number: MFCD12758183
CAS nummer: 1391449-79-8