7-Bromo-8-(trifluoromethyl)-1H-3,1-benzoxazine-2,4-dione 95%
Leverantör: Apollo Scientific
Varning
PC57425-250MG PC57425-1G PC57425-5G
APOSPC57425-250MGEA 2575 SEK
APOSPC57425-250MG APOSPC57425-1G APOSPC57425-5G
7-Bromo-8-(trifluoromethyl)-1H-3,1-benzoxazine-2,4-dione 95%
7-Bromo-8-(trifluoromethyl)-1H-3,1-benzoxazine-2,4-dione