2-Bromo-4-formylbenzoic acid
Leverantör: Apollo Scientific
Varning
OR400420-1G OR400420-250MG
APOSOR400420-1GEA 8225 SEK
APOSOR400420-1G APOSOR400420-250MG
2-Bromo-4-formylbenzoic acid
2-Bromo-4-formylbenzoic acid
Formel: C₈H₅BrO₃
MW: 229,03 g/mol
MDL Number: MFCD26855789
CAS nummer: 90001-44-8