(S)-tert-Butyl 5-methyl-1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide 97%
Leverantör: Apollo Scientific
Varning
Synonyms: (5S)-2,2-Dioxido-5-methyl-1,2,3-oxathiazolidine, N-BOC protected
OR400343-1G OR400343-5G OR400343-25G
APOSOR400343-1GEA 1600 SEK
APOSOR400343-1G APOSOR400343-5G APOSOR400343-25G
(S)-tert-Butyl 5-methyl-1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide 97%
(S)-tert-Butyl 5-methyl-1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide
Formel: C₈H₁₅NO₅S
MW: 237,27 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD09833247
CAS nummer: 396074-50-3