Ethylene glycol dimethacrylate (ethylene dimethacrylate) 95%
Leverantör: Apollo Scientific
Varning
Synonyms: Ethylene dimethacrylate, Ethane-1,2-diyl bis(2-methylacrylate)
OR936412-25G OR936412-100G
APOSOR936412-25GEA 415 SEK
APOSOR936412-25G APOSOR936412-100G
Ethylene glycol dimethacrylate (ethylene dimethacrylate) 95%
Ethylene glycol dimethacrylate (ethylene dimethacrylate)
Formel: C₁₀H₁₄O₄
Kokpunkt: 98 - 100 °C
Smältpunkt: −40 °C
Flampunkt: 104 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00008590
CAS nummer: 97-90-5