1,3-Dimethyl-1H-pyrazol-4-amine hydrochloride
Leverantör: Apollo Scientific
Varning
Synonyms: 4-Amino-1,3-dimethyl-1H-pyrazole hydrochloride
OR61322-100MG OR61322-250MG
APOSOR61322-100MGEA 245 SEK
APOSOR61322-100MG APOSOR61322-250MG
1,3-Dimethyl-1H-pyrazol-4-amine hydrochloride
1,3-Dimethyl-1H-pyrazol-4-amine hydrochloride
Formel: C₅H₁₀ClN₃
MW: 147,61 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD02253769
CAS nummer: 1147222-02-3