3-Bromo-2-chloro-5-iodobenzoic acid 95%
Leverantör: Apollo Scientific
Varning
OR61622-250MG OR61622-1G OR61622-5G
APOSOR61622-250MGEA 2250 SEK
APOSOR61622-250MG APOSOR61622-1G APOSOR61622-5G
3-Bromo-2-chloro-5-iodobenzoic acid 95%
3-Bromo-2-chloro-5-iodobenzoic acid
Formel: C₇H₃BrClIO₂
MW: 361.36 g/mol
MDL Number: MFCD26865670
CAS nummer: 860695-49-4