Methyl 3-bromo-5-chloro-2-iodobenzoate 95%
Leverantör: Apollo Scientific
Varning
OR61182-250MG OR61182-1G OR61182-5G
APOSOR61182-250MGEA 2250 SEK
APOSOR61182-250MG APOSOR61182-1G APOSOR61182-5G
Methyl 3-bromo-5-chloro-2-iodobenzoate 95%
Methyl 3-bromo-5-chloro-2-iodobenzoate
Formel: C₈H₅BrClIO₂
MW: 375.39 g/mol
MDL Number: MFCD07780742
CAS nummer: 933585-60-5