(±)-2,4-Difluoromandelic acid ≥97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: (±)-2,4-Difluorophenylglycolic acid
L10708.03 L10708.06
L10708.03EA 1550 SEK
L10708.03 L10708.06
(±)-2,4-Difluoromandelic acid ≥97%
(±)-2,4-Difluoromandelic acid
Purity: 97% CAS No.: 132741-30-1 Molecular Formula: C8H6F2O3 Formula Weight: 188.13 Melting Point: 122-124° MDL No.: MFCD00061296
Formel: C₈H₆F₂O₃
MW: 188.13 g/mol
Smältpunkt: 122…124 °C
Storage Temperature: Omgivande
MDL Number: MFCD00061296
CAS nummer: 132741-30-1