Gibberellinsyra ≥90%
Leverantör: Thermo Scientific
Varning
Synonyms: GA3, Gibberellin A3, (3S,3aS,4S,4aS,6S,8aR,8bR,11S)-6,11-Dihydroxy-3-methyl-12-methylene-2-oxo-4a,6-ethano-3,8b-prop-1-enoperhydroindeno[1,2-b]furan-4-carboxylic acid
A17843.03 A17843.06 A17843.14
A17843.03EA 935 SEK
A17843.03 A17843.06 A17843.14
Gibberellinsyra ≥90%
Gibberellinsyra
Purity: 90+% CAS No.: 77-06-5 Molecular Formula: C19H22O6 Formula Weight: 346.37 Melting Point: 222° dec. Beilstein Reference: 54346 Merck Reference: 14,4419 Optical Rotation: +78° (c=2 in methanol) MDL No.: MFCD00079329
Formel: C₁₉H₂₂O₆
MW: 346,38 g/mol
Smältpunkt: 222 °C (dec.)
Densitet: 1,386 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00079329
CAS nummer: 77-06-5
EINECS: 201-001-0
Merck Index: 14,4419