Gibberellinsyra 99%
Leverantör: Thermo Scientific
Varning
Synonyms: GA3, Gibberellin A3, (3S,3aS,4S,4aS,6S,8aR,8bR,11S)-6,11-Dihydroxy-3-methyl-12-methylene-2-oxo-4a,6-ethano-3,8b-prop-1-enoperhydroindeno[1,2-b]furan-4-carboxylic acid
410910010 410910050 410910250
ACRO410910010EA 1475 SEK
ACRO410910010 ACRO410910050 ACRO410910250
Gibberellinsyra 99%
Gibberellinsyra
Formel: C₁₉H₂₂O₆
MW: 346,38 g/mol
Smältpunkt: 227 °C (dec.)
Densitet: 1,386 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00079329
CAS nummer: 77-06-5
EINECS: 201-001-0
Merck Index: 13,04430
Order Now
Specifikation testresultat
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
Specific optical rotation +71° to +81° (20°C, 589 nm) (c=2, methanol)
Titration with NaOH ≥98.5 %