Sodium-5-(2-aminoethylamino)-1-naphthalenesulphonate hydrate ≥95% för molekylärbiologi

Leverantör: AAT BIOQUEST
Varning

Synonyms: Sodium N-(2-Aminoethyl)-5-naphthylamine-1-sulphonate, 5-(2-Aminoethylamino)-1-naphthalenesulphonic acid Sodium Salt, N-(2-Aminoethyl)-5-naphthylamine-1-sulphonic acid Sodium Salt, 1,5-EDANS sodium salt, EDANS, sodium salt

616 615
AATB616EA 16450 SEK
AATB616 AATB615
Sodium-5-(2-aminoethylamino)-1-naphthalenesulphonate hydrate ≥95% för molekylärbiologi
Sodium-5-(2-aminoethylamino)-1-naphthalenesulphonate hydrate

EDANS sodium salt is much more water-soluble than EDANS acid.


  • Useful for multiple applications

Formel: C₁₂H₁₄N₂O₃S
MW: 288.30 g/mol
Storage Temperature: Frys
MDL Number: MFCD00051474
CAS nummer: 100900-07-0
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e07330012b19671c95d43eb3b22c178a2a75f44&vid=c3d56bdb9772fa36efe073bdb8e253763bfa7e21&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR