5(6)-Tetramethylrhodamine isothiocyanate [5(6)-TRITC] 90
Leverantör: AAT BIOQUEST
Varning
Synonyms: Tetramethylrhodamine-5(6)-isothiocyanate, 5(6)-TRITC, MRITC, TRITC
409 410
AATB409EA 6825 SEK
AATB409 AATB410
5(6)-Tetramethylrhodamine isothiocyanate [5(6)-TRITC] 90
5(6)-Tetramethylrhodamine isothiocyanate [5(6)-TRITC]
5(6)-TRITC is an amino-reactive labeling reagent that is widely used in preparing bioconjugates of proteins and nucleic acids.
Formel: C₂₅H₂₁N₃O₃S
MW: 443.52 g/mol
MDL Number: MFCD00063517
CAS nummer: 95197-95-8
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a4608521e6bb3151793af30309e044bdac4417c&vid=d5a171979e5a3dbb9bbf12627ab5f5f10f77a5c1&prid=web&clid=SAPDAM