WST-8 50 mM, ReadiUse™ for assessing cell metabolism
Leverantör: AAT BIOQUEST
Varning
Synonyms: 2-(3-(2-Methoxy-4-nitrophenyl)-2-(4-nitrophenyl)-2H-tetrazol-3-ium-5-yl)-5-sulphobenzenesulphonate, sodium salt, 5-(2,4-Disulphophenyl)-3-(2-methoxy-4-nitrophenyl)-2-(4-nitrophenyl)-2H-tetrazolium inner salt monosodium salt
15707 15705 15706
AATB15707EA 13000 SEK
AATB15707 AATB15705 AATB15706
WST-8 50 mM, ReadiUse™ for assessing cell metabolism
WST-8
WST-8 (2-(2-methoxy-4-nitrophenyl)-3-(4-nitrophenyl)-5-(2,4-disulfophenyl)-2H-tetrazolium, monosodium salt) is a water-soluble tetrazolium salt used for assessing cell metabolic activity.
  • Improved sensitivity
  • Water soluble
  • High stability
  • Low cytotoxicity
Formel: C₂₀H₁₃N₆O₁₁S₂·Na
MW: 600.47 g/mol
MDL Number: MFCD09264687
CAS nummer: 193149-74-5
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fbeba4d47f5883428d6a35d6b51a87e0ae37d85&vid=df69d2ca107bba9f8352527730cc5ab502840edb&prid=web&clid=SAPDAM