Kinhydron ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: Benzoquinone-hydroquinone, p-Benzoquinone hydroquinone complex, 2,5-Cyclohexadiene-1,4-dione, compd. with 1,4-benzenediol (1:1)
A16681.14 A16681.22
A16681.14EA 935 SEK
A16681.14 A16681.22
Kinhydron ≥97%
Kinhydron
Formel: C₆H₄(OH)₂·C₆H₄O₂
MW: 218,21 g/mol
Smältpunkt: 168…171 °C
Densitet: 1,400
Storage Temperature: Omgivande
MDL Number: MFCD00010310
CAS nummer: 106-34-3
EINECS: 203-387-6
UN: 2811
ADR: 6.1,III
Merck Index: 14,8058