Natriumhydrid 57 - 63% dispersion i mineralolja
Leverantör: Thermo Scientific
Fara
13431.18 13431.36
13431.18EA 1025 SEK
13431.18 13431.36
Natriumhydrid 57 - 63% dispersion i mineralolja
Natriumhydrid
CAS No.: 7646-69-7 Molecular Formula: HNa Formula Weight: 24.00 Physical Form: Moist powder Density: 0.92 Sensitivity: Air and Moisture Sensitive Appearance: Off-white to gray Odour: Odourless Merck Reference: 14,8625 UN No.: 1427 MDL No.: MFCD00003471
Formel: HNa
MW: 24 g/mol
Densitet: 0,92
Storage Temperature: Omgivande
MDL Number: MFCD00003471
CAS nummer: 7646-69-7
EINECS: 231-587-3
UN: 1427
ADR: 4.3,I
Merck Index: 14,8625