Peridinin-Chlorophyll-Protein Complex fluorescent probe
Leverantör: AAT BIOQUEST
Varning
2540 2559
AATB2540EA 6500 SEK
AATB2540 AATB2559
Peridinin-Chlorophyll-Protein Complex fluorescent probe
Peridinin-Chlorophyll-Protein Complex
PerCP (Peridinin-chlorophyll-protein complex) is isolated from Dinophyceae sp.
  • High extinction coefficient
  • High quantum efficiency
  • Large Stokes shift
ADR: 5,II