LLE High Recovery Vial Racks and Vials
Leverantör: EPREP
01-03126
EPRE01-03126EA 615 SEK
EPRE01-03126
LLE High Recovery Vial Racks and Vials
Sample Preparation Instruments
ePrep 20 and 50 ml high recovery PP vials are ideal for syringe LLE techniques (Patent Pending) on the ePrep Sample Preparation Workstation.