Cole-Parmer® Stuart DR-200 Rotating Shaker System Drives
Leverantör: Cole Parmer
51600-06
51600-92
51600-93
51600-91
51600-94
51600-95
51600-96
51600-97
51600-98
51600-99
STUA51600-06EA
20400
SEK
STUA51600-06
STUA51600-92
STUA51600-93
STUA51600-91
STUA51600-94
STUA51600-95
STUA51600-96
STUA51600-97
STUA51600-98
STUA51600-99
Cole-Parmer® Stuart DR-200 Rotating Shaker System Drives
Skakapparater
Skakapparater, roterande/vaggande rörelse
SPECIFIKATIONER
Environmentally Preferable | ||
Hastighetsområde | 6 - 60 min⁻¹ | |
B×D×H | 650×250×250 mm | |
Elektrisk | 230 VAC, 50 Hz | |
Max. load | 3 kg | |
Timer | 10 - 60 min |