Caprylic/capric triglyceride
Leverantör: Spectrum Chemical
Synonyms: Miglyol 810, Triglyceride of Coconut Oil, Neobee M5, Glycerides, mixed Decanoyl and Octanoyl
C3465-200LTBL C3465-2.5LT C3465-500ML
SPCMC3465-200LTBLEA 102600 SEK
SPCMC3465-200LTBL SPCMC3465-2.5LT SPCMC3465-500ML
Caprylic/capric triglyceride
Caprylic/capric triglyceride
Caprylic/Capric Triglyceride is a medium-chain triglyceride used as a component of ointments, creams, and liquid emulsions.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formel: C₂₁H₃₉O₆
MW: 387,5 g/mol
Storage Temperature: Omgivande
CAS nummer: 73398-61-5
Order Now
Specifikation testresultat
Saponification Value 325 - 360
Specific Gravity 0.94 - 0.96