Polyethylene glycol monostearate 40 NF Type II
Leverantör: Spectrum Chemical
Synonyms: Polyoxyethylene stearate, Polyoxyl stearate
P1159-500GM P1159-12KG P1159-2.5KG P1159-45KGBL P1159-113KGBL
SPCMP1159-500GMEA 3600 SEK
SPCMP1159-500GM SPCMP1159-12KG SPCMP1159-2.5KG SPCMP1159-45KGBL SPCMP1159-113KGBL
Polyethylene glycol monostearate 40 NF Type II
Polyethylene glycol monostearate
Polyoxyl 40 Stearate, Type II, NF is commonly used in pharmaceutical formulations as an emulsifier and a surfactant.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formel: HO(CH₂CH₂O)nOCC₁₇H₃₅
Storage Temperature: Omgivande
MDL Number: MFCD00148007
CAS nummer: 9004-99-3
Order Now
Specifikation testresultat
Stearic Acid and Palmitic Acid 96.0% min.-
Stearic Acid 90.0 - 99.0 %
Free Polyethylene Glycols 17 - 27 %
Alkalinity To Pass Test
Hydroxyl Value 25 - 40