6-O-Palmitoyl-L-ascorbic acid ≥95.0% NF
Leverantör: Spectrum Chemical
Synonyms: L-Ascorbic acid 6-palmitate, Palmitoyl-L-ascorbic acid, Ascorbyl Palmitate, (S)-2-((R)-3,4-Dihydroxy-5-oxo-2,5-dihydrofuran-2-yl)-2-hydroxyethyl palmitate, L(+)-Ascorbyl palmitate
A1372-25KGBL A1372-50KGBL A1372-5KGBL
SPCMA1372-25KGBLEA 0 SEK
SPCMA1372-25KGBL SPCMA1372-50KGBL SPCMA1372-5KGBL
6-O-Palmitoyl-L-ascorbic acid ≥95.0% NF
6-O-Palmitoyl-L-ascorbic acid
Ascorbyl Palmitate, NF is often used to promote flow of tableting powders and to prevent their sticking in tablet presses.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formel: C₂₂H₃₈O₇
MW: 414.54 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00005377
CAS nummer: 137-66-6
Order Now
Specifikation testresultat
Assay (Dried Basis) 95.0 - 100.5%
Specific Rotation [A]D25 +21° to +24°
Loss On Drying max. 2.0 %
Residue On Ignition max. 0.1 %
Heavy Metals max. 10 ppm