EDTA trisodium salt ≥99%
Leverantör: Spectrum Chemical
E1046-12KG E1046-22.5KGBL E1046-50KGBL
SPCME1046-12KGEA 0 SEK
SPCME1046-12KG SPCME1046-22.5KGBL SPCME1046-50KGBL
EDTA trisodium salt ≥99%
EDTA trisodium salt
Ethylenediaminetetraacetic Acid Trisodium Salt is a colourless aminopolycarboxylic acid that is water soluble. It is typically used to dissolve lime scale. It is valuable because of its role as a six toothed ligand and chelating agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Formel: C₁₀H₁₃N₂Na₃O₈
MW: 358,19 g/mol
Smältpunkt: > 300 °C
Densitet: 0,7215 g/cm³ (20 °C)
Storage Temperature: Omgivande
CAS nummer: 150-38-9
EINECS: 205-758-8
Merck Index: 12,03558
Order Now
Specifikation testresultat
Assay 99% Min.