D-(-)-Mannitol ≥96%, pulver USP
Leverantör: Spectrum Chemical
Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
MA165-2.5KG MA165-500GM
SPCMMA165-2.5KGEA 12275 SEK
SPCMMA165-2.5KG SPCMMA165-500GM
D-(-)-Mannitol ≥96%, pulver USP
D (-)-Mannitol
Mannitol, Powder, USP.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel: HOH₂C(CH(OH))₄CH₂OH
Kokpunkt: 290…295 °C (4 hPa)
Smältpunkt: 164…169 °C
Densitet: 1,49 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00064287
CAS nummer: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Assay 96% Min.