Tributyrin ≥97%
Leverantör: Apollo Scientific
Synonyms:
Tributyrin, Glyceryl tributyrate, Glycerol tributyrate, Propane-1,2,3-triyl tributyrate
OR62132-25G
OR62132-100G
OR62132-500G
APOSOR62132-25GEA
380
SEK
APOSOR62132-25G
APOSOR62132-100G
APOSOR62132-500G
Tributyrin ≥97%
Tributyrin
Formel:
C₁₅H₂₆O₆ MW: 302,37 g/mol Kokpunkt: 129-131 °C/0,03mm Smältpunkt: -75 °C Densitet: 1,032 Flampunkt: 174 °C |
MDL Number:
MFCD00009392 CAS nummer: 60-01-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...