Dimethyl succinate zS, Sigma-Aldrich®
Leverantör: Merck
Varning
Synonyms: DBE-4 dibasic ester, Succinic acid dimethyl ester, 1,4-Butanedioic acid dimethyl ester
MQ200
8.44002.0100 8.44002.1000
8.44002.0100EA 320 SEK
8.44002.0100 8.44002.1000
Dimethyl succinate zS, Sigma-Aldrich®
Dimethyl succinate
Formel: C₆H₁₀O₄
MW: 146.14 g/mol
Kokpunkt: 200 °C (1013 hPa)
Smältpunkt: 18…19 °C
Densitet: 1.117 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00008466
CAS nummer: 106-65-0
EINECS: 203-419-9
Merck Index: 12,09038