Normal flow filter cartridges, ULTA Pure HC
Leverantör: Cytiva
12-4100-93 12-4100-94 12-4100-97
12-4100-93EA 3925 SEK
12-4100-93 12-4100-94 12-4100-97
Normal flow filter cartridges, ULTA Pure HC
Filter Filterpatroner
Sterilization-grade filter cartridges with a polyethersulfone prefilter for long life and high capacity filtration.
  • Sterilizing grade 0,2 µm PES (polyethersulfone) membrane with prefiltration layer
  • Standard 5" filter cartridge.
  • Optimized membrane configuration allows up to ten times the throughput to blockage.
  • Integral prefilter layer can condense filter trains for greater processing economy.
  • Fully validated and integrity testable membrane for assurance of sterility.
  • Low adsorption of proteins for minimal loss of expensive pharmaceutical product.