DL-α-Tocopherol (Vitamin E) 96,0 USP
Leverantör: Spectrum Chemical
Synonyms: (±)-α-Tocopherol, Vitamin E Alcohol, all-rac-α-Tocopherol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-ol, DL-alpha-Tocopherol, Vitamin E
VI135-1LTGL VI135-100ML VI135-500ML
SPCMVI135-1LTGLEA 23650 SEK
SPCMVI135-1LTGL SPCMVI135-100ML SPCMVI135-500ML
DL-α-Tocopherol (Vitamin E) 96,0 USP
DL-α-Tocopherol (Vitamin E)
Vitamin E, 1 mg = 0.74 alpha-tocopherol equivalents, USP. 
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel: C₂₉H₅₀O₂
MW: 430.71 g/mol
Densitet: 0.950
Storage Temperature: Kylskåp
MDL Number: MFCD00006848
CAS nummer: 10191-41-0
Order Now
Specifikation testresultat
Assay (C19H50O₂) 96.0 - 102.0 %
Acidity To Pass Test
Identification To Pass Test