Nikotinamid USP
Leverantör: Spectrum Chemical
Varning
Synonyms: 3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
NI105-1KG NI105-25KGBL
SPCMNI105-1KGEA 3425 SEK
SPCMNI105-1KG SPCMNI105-25KGBL
Nikotinamid USP
Nikotinamid
Niacinamide, USP is a form of vitamin B3. 
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel: C₆H₆N₂O
MW: 122,13 g/mol
Kokpunkt: 334 °C (1013 hPa)
Smältpunkt: 127…128 °C
Densitet: 1,4463 g/cm³ (20 °C)
Flampunkt: 182 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006395
CAS nummer: 98-92-0
EINECS: 202-713-4
Merck Index: 12,06574
Order Now
Specifikation testresultat
Assay 98.5% Min.