Natriumalginat NF
Leverantör: Spectrum Chemical
S1118-2.5KG S1118-500GM
SPCMS1118-2.5KGEA 9300 SEK
SPCMS1118-2.5KG SPCMS1118-500GM
Natriumalginat NF
Natriumalginat
Sodium Alginate, NF is a gum that has no discernible flavor and is used in the food and pharmaceutical industries as an emulsifier and to increase viscosity. All Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Storage Temperature: Omgivande
MDL Number: MFCD00081310
CAS nummer: 9005-38-3
Order Now
Specifikation testresultat
Assay 90.8% Min.