Propyl-4-hydroxibensoat ≥98% Ph. Eur., BP, NF
Leverantör: Spectrum Chemical
Varning
PR133-500GM PR133-100GM PR133-12KG PR133-2.5KG PR133-25KGBL
SPCMPR133-500GMEA 2750 SEK
SPCMPR133-500GM SPCMPR133-100GM SPCMPR133-12KG SPCMPR133-2.5KG SPCMPR133-25KGBL
Propyl-4-hydroxibensoat ≥98% Ph. Eur., BP, NF
Propyl-4-hydroxibensoat
Propylparaben, NF, BP, EP is used as an antimicrobial preservative. All Spectrum Chemical NF, BP, EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel: C₁₀H₁₂O₃
Kokpunkt: 292 °C (1013 hPa)
Smältpunkt: 95 °C
Densitet: 1,28 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002354
CAS nummer: 94-13-3
EINECS: 202-307-7
Merck Index: 13,07958
Order Now
Specifikation testresultat
Assay 98% Min.