L (+)-Serin ≥98.5% FCC
Leverantör: Spectrum Chemical
Synonyms: H-Ser-OH, (S)-2-Amino-3-hydroxypropanoic acid, Serine, (S)-(+)-Serine
S1021-100GM S1021-1KG S1021-25GM
SPCMS1021-100GMEA 3075 SEK
SPCMS1021-100GM SPCMS1021-1KG SPCMS1021-25GM
L (+)-Serin ≥98.5% FCC
L (+)-Serin
L-Serine, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel: C₃H₇NO₃
Kokpunkt: 259 °C (1013 hPa)
Smältpunkt: 215…225 °C
Densitet: 1,6 g/cm³ (22 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00064224
CAS nummer: 56-45-1
EINECS: 200-274-3
Merck Index: 14,00846
Order Now
Specifikation testresultat
Assay 98.5% Min.